Läs mer
9-Ethyl-9H-carbazole-3-carboxylic Acid, TRC
Produktkod. 30380832
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30380832 5 mg
30377423 50 mg
30371489 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30380832 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9039055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57102-98-4
Molekylformel C15H13NO2
Molekylvikt (g/mol) 239.27
LEDER CCn1c2ccccc2c3cc(ccc13)C(=O)O
Specifikationer
Kemiskt namn eller material 9-Ethyl-9H-carbazole-3-carboxylic Acid
Kvantitet 5 mg
InChI formel InChI=1S/C15H13NO2/c1-2-16-13-6-4-3-5-11(13)12-9-10(15(17)18)7-8-14(12)16/h3-9H,2H2,1H3,(H,17,18)
Formel vikt 239.0946