Läs mer
9-Fluoro-11beta,17-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-21-yl Propanoate (Betamethasone 21-Propionate), Mikromol™
Produktkod. 30051794
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051794 25 mg
18030660 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051794 Leverantör Mikromol Leverantörsnummer MM0202.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
9-Fluoro-11beta,17-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-21-yl Propanoate (Betamethasone 21-Propionate)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Betamethasone 21-Propionate
CAS 75883-07-7
Analyt- eller komponentnamn Betamethasone 21-Propionate
Frakt skick Room Temperature
Molekylformel C25 H33 F O6
Certifikat och efterlevnad ISO 17025
Synonym Betamethasone 21-Propionate, 9-Fluoro-11beta,17-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-21-yl Propanoate, Clobetasol Propionate Imp. K (EP), Betamethasone Dipropionate Imp. C (Pharmeuropa), Betamethasone Dipropionate Related Compound C, Betamethasone Dipropionate Impurity C, Clobetasol Propionate Impurity K
InChI formel InChI=1S/C25H33FO6/c1-5-21(30)32-13-20(29)25(31)14(2)10-18-17-7-6-15-11-16(27)8-9-22(15,3)24(17,26)19(28)12-23(18,25)4/h8-9,11,14,17-19,28,31H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1
LEDER CCC(=O)OCC(=O)[C@@]1(O)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
Molekylvikt (g/mol) 448.52
Kvantitet 25 mg
Formel vikt 448.2261 g/mol