Läs mer
9-Fluoro-11beta-hydroxy-16alpha,17-(1-methylethylidenedioxy)-3,20-dioxopregna-1,4-dien-21-yl Acetate (21-Acetate Triamcinolone Acetonide), Mikromol™
Produktkod. 18072790
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18072790 100 mg
18043100 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18072790 Leverantör Mikromol Leverantörsnummer MM0198.09
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
9-Fluoro-11beta-hydroxy-16alpha,17-(1-methylethylidenedioxy)-3,20-dioxopregna-1,4-dien-21-yl Acetate (21-Acetate Triamcinolone Acetonide)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Triamcinolone Acetonide 21-Acetate
CAS 7-3-3870
Analyt- eller komponentnamn Triamcinolone Acetonide 21-Acetate
Frakt skick Room Temperature
Molekylformel C26 H33 F O7
Certifikat och efterlevnad ISO 17025
Synonym (11beta,16alpha)-21-(acetyloxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregna-1,4-diene-3,20-dione, 9-Fluoro-11β,16α,17,21-tetrahydroxy-pregna-1,4-diene-3,20-dione 21-acetate cyclic 16,17-acetal with acetone, 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregna-1,4-diene-3,20-dione deriv. Triamcinolone acetonide 21-acetate, Triamcinolone Acetonide Impurity F
InChI formel InChI=1S/C26H33FO7/c1-14(28)32-13-20(31)26-21(33-22(2,3)34-26)11-18-17-7-6-15-10-16(29)8-9-23(15,4)25(17,27)19(30)12-24(18,26)5/h8-10,17-19,21,30H,6-7,11-13H2,1-5H3/t17-,18-,19-,21+,23-,24-,25-,26+/m0/s1
LEDER CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C
IUPAC-namn (11β,16α)-21-(Acetyloxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregna-1,4-diene-3,20-dione
Molekylvikt (g/mol) 476.53
Kvantitet 100 mg
Formel vikt 476.221 g/mol