Learn More
9-Methylacridine, TRC
Product Code. 30132630
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132630 10 g 10g
30009290 100 mg 100mg
30008524 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132630 Supplier Toronto Research Chemicals Supplier No. TRCM28505010G
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 611-64-3
Molecular Formula C14 H11 N
Molecular Weight (g/mol) 193.24
Synonym 9-Methylacridine, Carbamazepine Imp. B (EP)
IUPAC Name 9-methylacridine
SMILES Cc1c2ccccc2nc3ccccc13
Specifications
Quantity 10 g
InChI Formula InChI=1S/C14H11N/c1-10-11-6-2-4-8-13(11)15-14-9-5-3-7-12(10)14/h2-9H,1H3
Formula Weight 193.0891
Chemical Name or Material 9-Methylacridine