Learn More
9-Methylanthracene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.
Solubility
Soluble in water (0,0003 mg/L at 25°C) and toluene. Slightly soluble in benzene.
Notes
Mechanical exhaust required. When not in use, tightly seal the container and store in a dry, cool place.
9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.
Solubility
Soluble in water (0,0003 mg/L at 25°C) and toluene. Slightly soluble in benzene.
Notes
Mechanical exhaust required. When not in use, tightly seal the container and store in a dry, cool place.
Specifications
Melting Point | 77°C to 79°C |
Density | 1.066 |
Boiling Point | 196°C to 197°C (16 mmHg) |
Quantity | 1 g |
UN Number | UN3077 |
Beilstein | 1907463 |
Solubility Information | Soluble in water (0,0003mg/L at 25°C) and toluene. Slightly soluble in benzene. |
Formula Weight | 192.26 |
Percent Purity | 99% |
Chemical Name or Material | 9-Methylanthracene |