Learn More
9-Methylanthracene, 99%, Thermo Scientific Chemicals
Catalog Number p-4509807
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
779-02-2
C15H12
192.26
MFCD00001261
CPGPAVAKSZHMBP-UHFFFAOYSA-N
anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard
13068
9-methylanthracene
CC1=C2C=CC=CC2=CC2=CC=CC=C12
779-02-2
C15H12
192.26
MFCD00001261
CPGPAVAKSZHMBP-UHFFFAOYSA-N
anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard
13068
9-methylanthracene
CC1=C2C=CC=CC2=CC2=CC=CC=C12
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 76.0°C to 79.0°C
Color Orange to Yellow
Density 1.0660g/mL
Boiling Point 196.0°C to 197.0°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 05,II,586
Specific Gravity 1.066
Formula Weight 192.26
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 9-Methylanthracene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 9-Methylanthracene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.