Läs mer
9-Octadecenenitrile, TRC
Produktkod. 30132494
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132494 250 mg
30134180 100 mg
17992295 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132494 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO724140250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 112-91-4
Molekylformel C18 H33 N
Molekylvikt (g/mol) 263.4613
Synonym Oleic Acid Nitrile, Oleic Nitrile, Oleoylnitrile, Oleyl Nitrile, Arneel O, (9Z)-9-Octadecenenitrile, Oleonitrile (6CI,7CI,8CI)
IUPAC-namn (Z)-octadec-9-enenitrile
LEDER CCCCCCCC\C=C/CCCCCCCC#N
Specifikationer
Kemiskt namn eller material 9-Octadecenenitrile
Anteckningar om renhetsgrad GC
Kvantitet 250 mg
InChI formel InChI=1S/C18H33N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10H,2-8,11-17H2,1H3/b10-9-
Formel vikt 263.2613
Procent renhet >95%