Learn More
9-Vinylanthracene, 98%, BLD Pharm™
Product Code. 30944208
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30944208 5 g 5g
30938415 1 g 1g
30930676 25 g 25g
30937005 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30944208 Supplier BLD Pharm Supplier No. BD1118695g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 2444-68-0
Molecular Formula C16H12
MDL Number MFCD00003576
Quantity 5 g
SMILES C=CC1=C2C=CC=CC2=CC3=CC=CC=C13
Molecular Weight (g/mol) 204.27
Percent Purity 98%
Chemical Name or Material 9-Vinylanthracene