Learn More
Uvex™ 9188 Cybric Safety Spectacles
Cybric spectacles offer a large field of vision and individual adjustment capabilities for reliable protection and comfort.
Brand:  Uvex™ 9188075
Additional Details : Weight : 0.05400kg
 View more versions of this product
Product Code. 15396052
  • 149.00 SEK / Each
In Stock
to see stock.
Description
Description
  • Spectacles feature a large field of vision and individual adjustment capabilities for reliable protection and comfort
  • Certified according to EN 166 (personal eye protection) and EN 170 (UV filters)
  • Quattroflex side arms with four comfort cushions to ensure a comfortable, pressure-free fit around the ear
  • Inclination of the side arms can be adjusted between multiple levels, allowing them to be adapted individually to all facial shapes
  • Supravision sapphire coating is extremely chemical-resistant and scratch-resistant on both sides
  • Reliable UV 400 protection
Specifications
Specifications
EN 166:2001, EN 170:2002
Black, Orange
Supravision Sapphire
UV 400
Clear
9188 Cybric
Polycarbonate