Learn More
Uvex™ 9188 Cybric Safety Spectacles
Cybric spectacles offer a large field of vision and individual adjustment capabilities for reliable protection and comfort.
149.00 SEK - 177.00 SEK
Specifications
Certifications/Compliance (Europe) | EN 166:2001, EN 170:2002 |
---|---|
Frame Color | Black, Orange |
Lens Coating | Supravision Sapphire |
Lens Material | Polycarbonate |
Model | 9188 Cybric |
Product Code | Brand | Lens Tint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Lens Tint | Price | Quantity & Availability | |||||
15396052
|
Uvex™
9188075 |
Clear |
149.00 SEK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
10064442
|
Uvex™
9188076 |
Gray |
177.00 SEK
Each |
Estimated Shipment: 26-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
- Spectacles feature a large field of vision and individual adjustment capabilities for reliable protection and comfort
- Certified according to EN 166 (personal eye protection) and EN 170 (UV filters)
- Quattroflex side arms with four comfort cushions to ensure a comfortable, pressure-free fit around the ear
- Inclination of the side arms can be adjusted between multiple levels, allowing them to be adapted individually to all facial shapes
- Supravision sapphire coating is extremely chemical-resistant and scratch-resistant on both sides
- Reliable UV 400 protection
Specifications
EN 166:2001, EN 170:2002 | |
Supravision Sapphire | |
9188 Cybric |
Black, Orange | |
Polycarbonate | |
UV 400 |