Läs mer
uvex 9192 Pheos glasögon
Handla allt Uvex Produkter
Linsbeläggning:
Anti-dimma, Reptålig
Anti-fog
Anti-fog, Scratch-resistant
Linsfärg:
Clear
Gray
Klart
Beskrivning
- Soft, non-slip arm end prevents pressure points for enhanced comfort
- Duo-spherical lenses provide a large field of vision
- Design is extremely compatible with ear defenders, helmets, and bump caps
- Eye Plus Protection provides additional eyebrow protection with a bevelled edge in the upper area
- Easily add a headband and frame when working at heights or in confined spaces
Supravision Excellence Coating
- Interior: Anti-fog
- Exterior: Scratch-resistant and chemical-resistant
- Lenses are easy to clean and less susceptible to soiling
Supravision Plus Coating
- Inside: Anti-fog and scratch-resistant
- Outside: Anti-fog and scratch-resistant
- Suitable for environments with frequent temperature and humidity fluctuations
Specifikationer
Specifikationer
| Certifikat och efterlevnad | EN 166:2001, EN 170:2002 |
| Linstyp | Duo-Spherical |
| Linsbeläggning | Anti-fog |
| Linsfärg | Clear |
| Produkttyp | Ergonomic Safety Glasses |
| Ram färg | Black, Orange |
| Modell | 9192 Pheos |
| Material | Polycarbonate |
| Disponibel | Reusable |
| Linsmaterial | Polycarbonate |
| Visa mer |