Läs mer
Uvex™ 9195 Skyper Safety Spectacles
Handla allt Uvex Produkter
:
Brown
Beskrivning
- Extremely lightweight safety spectacles with a modern design
- Supravision sapphire coating is scratch-resistant and chemical-resistant
- Individual temple inclination, and length adjustment for a personalized fit
- Simple lens replacement
- Soft temples
Specifikationer
Specifikationer
| Linstyp | Supravision Sapphire |
| Certifieringar/efterlevnad (Europa) | EN 166:2001, EN 172:1994 + A1:2000 + A2:2001 |
| Linsfärg | Brown |
| Ram färg | Black |
| Modell | 9195 Sykper |
| Linsmaterial | Polycarbonate |
| UV-skydd | UV 400 |