Learn More
Uvex 94908 Comfortable Climate Insole For Uvex G2 - Width 11
Comfortable climatic insole with antistatic properties for Uvex 1 G2
Brand:  Uvex 9490836
Additional Details : Weight : 0.04900kg
 View more versions of this product
Product Code. 17227431
  • 204.00 SEK / 1 pair
Estimated Shipment: 26-10-2023
to see stock.
Add to basket
Description
Description
  • Full-surface foot shock absorption
  • Highly breathable, moisture-absorbent and quickly re-drying material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No: 9490.8
  • Sizes: 35 to 52
Specifications
Specifications
Black
Microvelour
No
Standard width
No
Uvex G2
36
Unisex