Learn More
Uvex 95282 Medium Tune-Up Insole For Uvex 1 and 2 - Width 11
Tune-Up antistatic insole for reducing fatigue and supporting arch
Brand:  Uvex 9528237
Additional Details : Weight : 0.03500kg
 View more versions of this product
Product Code. 17218021
  • 251.00 SEK / 1 pair
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Description
Description
Uvex Tune-Up Comfortable Medium Insole 95282
  • Antistatic insole concept for use in safety footwear
  • Designed to reduce fatigue by increasing wearer comfort and enhancing ergonomics by optimum support of arch of feet
  • Good cushioning
  • Relief of pressure marks
  • Non-irritating, comfortable surface structure
  • Breathable and absorbs moisture
  • Antibacterial
  • Antistatic
  • Useable for ESD-footwear
  • Uvex 1, Uvex 2, Uvex 1 Duo, Uvex Xenova™ Light
  • Article No. 95282
  • Standard EN ISO 20345:2011 S1 SRC
  • Sizes: 35 to 52
Specifications
Specifications
Black, White
Microvelour
No
Standard width
No
Uvex 1 and 2
37
Unisex
Yes