Learn More
Uvex 95285 Medium Tune-Up Insole For Uvex 1 Sport - Width 11
Tune-Up antistatic insole for reducing fatigue and supporting arch
Brand:  Uvex 9528544
Additional Details : Weight : 0.13600kg
 View more versions of this product
Product Code. 17228081
  • 223.00 SEK / 1 pair
Estimated Shipment: 29-06-2023
to see stock.
Add to basket
Description
Description
Uvex Tune-Up Comfortable Medium Insole 95285
  • Antistatic insole concept for use in safety footwear
  • Designed to reduce fatigue by increasing wearer comfort and enhancing ergonomics by optimum support of arch of feet
  • Good cushioning
  • Relief of pressure marks
  • Non-irritating, comfortable surface structure
  • Breathable and absorbs moisture
  • Antibacterial
  • Antistatic
  • Useable for ESD-footwear
  • Uvex 1 Sport
  • Article No. 95285
  • Standard EN ISO 20345:2011
  • Sizes: 35 to 52
Specifications
Specifications
Black, White
Microvelour
No
Standard width
No
Uvex 1 Sport
44
Unisex