missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ 9770 Pheos ES Unventilated Safety Helmets

Lightweight, unventilated helmets in a sporty design ideal for use in the electrician sector with low voltage systems. Features a shortened shield with a conventional interior for easy adjustment.

Brand:  Uvex™ 9770521

94.00 SEK valid until 2023-05-26
Use promo code "20831" to get your promotional price.



Additional Details : Weight : 0.39000kg

 View more versions of this product

Product Code. 17229111

  • 141.00 SEK / Each

Estimated Shipment: 28-06-2023
to see stock.
Add to basket

Description

Description

  • Lightweight, unventilated helmet in a sporty design
  • Completely closed for use in the electrician sector with low voltage systems
  • Includes an adapter system for visors, helmet caps, and helmet lamps
  • Conventional interior for easy adjustment
  • Shortened shield
Specifications

Specifications

EN 397:2012, EN 50365:2002
Low Voltage Systems
HDPE
Electrically Insulating Helmet
Not Ventilated
Blue
Adapter System for Uvex™ Visors, Helmet Capsules, and Helmet Lamps
9770 Pheos ES
52 to 61 cm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers