Learn More
uvex 9906 Pheos Replacement Visor Lens SLB2
Product Code. 17230282 Shop All Uvex Products
Change view
Click to view available options
SLB2
Product Code.
17230282 SLB2
1 options
Product Code. 17230282 Supplier uvex Supplier No. 9906017
  • ONLINE EXCLUSIVE PRICE
    997.00 SEK
    942.98 SEK / Paket med 1
    Save 54.02 SEK
    5% Av
Beräknad leverans: 04-12-2026
Replacement visor lens for pheos series safety helmets is compatible with model 9906.002 (mechanical lever arms), or model 9906.003 (self-locking magnetic catches).
  • Replacement lens for uvex pheos model 9906.002 (mechanical lever arms) or model 9906.003 (self-locking magnetic catches)
  • Lightweight, robust face shield for uvex pheos industrial safety helmet ES-WR / BS-WR and uvex pheos alpine (visor adapter 9790.067 required)
  • Labeling: 2C-1,2 W1 B 8-2-0 9 KN CE 0196
  • Features supravision excellence coating
  • Arc flash class 2 with chin protection
Supravision Excellence Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant and chemical-resistant
  • Easy to clean, and less susceptible to soiling
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001, EN 170:2002
For Use With (Equipment) Pheos 9906.002, 9906.003
Includes Chin Protection
Lens Tint Blue
Material Polycarbonate