missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ 9954 Fabric Visor Pouch
Click to view available options
Black
Description
- Designed for safe storage and transportation of Uvex™ Visors and Safety Helmets
Specifications
Color | Black |
Product Type | Storage Case |
For Use With (Equipment) | Uvex™ Visors and Safety Helmets |
For Use With (Application) | Storage and Transportation |
Material | Microfiber |
Disposable | Reusable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'