Läs mer
9Alpha-Fluoroprednisolone 17-Valerate, TRC
Produktkod. 30133080
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133080 100 mg
30132726 25 mg
30011785 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133080 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF595615100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2802-10-0
Molekylformel C26 H35 F O6
Molekylvikt (g/mol) 462.55
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11,21-dihydroxy-17-[(1-oxopentyl)oxy]-, (11β)- (9CI), Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-, 17-valerate (7CI,8CI), 9α-Fluoroprednisolone 17-valerate, Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-dihydroxy-, 17-valerate, Betamethasone Valerate Imp. I (EP)
IUPAC-namn [(8S,9R,10S,11S,13S,14S,17R)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] pentanoate
LEDER CCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO
Specifikationer
Kemiskt namn eller material 9alpha-Fluoroprednisolone 17-Valerate
Kvantitet 100 mg
InChI formel InChI=1S/C26H35FO6/c1-4-5-6-22(32)33-25(21(31)15-28)12-10-18-19-8-7-16-13-17(29)9-11-23(16,2)26(19,27)20(30)14-24(18,25)3/h9,11,13,18-20,28,30H,4-8,10,12,14-15H2,1-3H3/t18-,19-,20-,23-,24-,25-,26-/m0/s1
Formel vikt 462.2418