Läs mer
9beta,11beta-Epoxy-17,21-dihydroxy-16beta-methylpregna-1,4-diene-3,20-dione, TRC
Produktkod. 17960415
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17960415 250 mg
17991885 1 g
17994565 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17960415 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE589175250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 981-34-0
Molekylformel C22 H28 O5
Molekylvikt (g/mol) 372.45
Synonym Pregna-1,4-diene-3,20-dione, 9,11-epoxy-17,21-dihydroxy-16-methyl-, (9β,11β,16β)-, 9β-Pregna-1,4-diene-3,20-dione, 9,11β-epoxy-17,21-dihydroxy-16β-methyl- (6CI,7CI,8CI), 9,11-Epoxy-9H-cyclopenta[a]phenanthrene, pregna-1,4-diene-3,20-dione deriv., 9,11β-Epoxy-17,21-dihydroxy-16β-methyl-9β-pregna-1,4-diene-3,20-dione, Betamethasone 9,11-epoxide
IUPAC-namn 9,11β-Epoxy-17,21-dihydroxy-16β-methyl-9β-pregna-1,4-diene-3,20-dione
LEDER C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]2(C)[C@@]1(O)C(=O)CO
Specifikationer
Kemiskt namn eller material 9,11beta-Epoxy-17,21-dihydroxy-16beta-methyl-9beta-pregna-1,4-diene-3,20-dione
Kvantitet 250 mg
InChI formel InChI=1S/C22H28O5/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)22(15)18(27-22)10-20(16,3)21(12,26)17(25)11-23/h6-7,9,12,15-16,18,23,26H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,18-,19-,20-,21-,22+/m0/s1
Formel vikt 372.1937