Läs mer
(9E)-Lumefantrine, TRC
Produktkod. 30477336
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477336 25 mg
30470284 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30477336 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL47400525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1456736-33-6
Molekylformel C30 H32 Cl3 N O
Molekylvikt (g/mol) 528.94
Synonym (9E)-2,7-Dichloro-9-[(4-Chlorophenyl)Methylene]-α-[(Dibutylamino)Methyl]-9H-Fluorene-4-Methanol
IUPAC-namn 2-(dibutylamino)-1-[(9E)-2,7-dichloro-9-[(4-chlorophenyl)methylidene]fluoren-4-yl]ethanol
LEDER CCCCN(CCCC)CC(O)c1cc(Cl)cc2\C(=C\c3ccc(Cl)cc3)\c4cc(Cl)ccc4c12
Specifikationer
Kemiskt namn eller material (9E)-Lumefantrine
Kvantitet 25 mg
InChI formel InChI=1S/C30H32Cl3NO/c1-3-5-13-34(14-6-4-2)19-29(35)28-18-23(33)17-27-25(15-20-7-9-21(31)10-8-20)26-16-22(32)11-12-24(26)30(27)28/h7-12,15-18,29,35H,3-6,13-14,19H2,1-2H3/b25-15+
Formel vikt 527.155