Learn More
Thermo Scientific™ 9mm Non-Assembled Clear Screw Thread Wide Opening Autosampler Vial Kits
Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 clear glass vials and 100 screw thread caps with pre-assembled septa.
Brand:  Thermo Scientific™ 60180-694
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13141773
  • 528.58 SEK / Pack of 100
Estimated Shipment date
from Supplier 09-07-2019
Add to basket
Description
Description
  • Convenience Kits holds vials and PINK closures in a two-compartment benchtop storage tray
  • Kits include 100 vials and 100 PINK closures
  • All items remain handy and dust-free
Specifications
Specifications
Red PTFE/White Silicone Screw
Screw
Autosampler Vials
100
Clear
1.5 mL
9mm
9mm Conv. Kit, clear vial, Red PTFE/White Silicone
Yes
Assembled Vial Kit
Glass
Videos
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.