Learn More
Thermo Scientific™ 9mm Non-Assembled Clear Screw Thread Wide Opening Autosampler Vial Kits
Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 clear glass vials and 100 screw thread caps with pre-assembled septa.
Brand:  Thermo Scientific™ VAGK
Additional Details : Weight : 0.28952kg
 View more versions of this product
Product Code. 13296419
  • 281.75 SEK / Pack of 100
Remote Stock
(10 days availability): 1
Add to basket
Specifications
Specifications
AVCS Convenience Kit, for Varian GC Autosampler
100
Glass
Yes
Clear
Videos
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.