Learn More
Thermo Scientific™ 9mm Non-Assembled Clear Screw Thread Wide Opening Autosampler Vial Kits
Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 clear glass vials and 100 screw thread caps with pre-assembled septa.
Brand:  Thermo Scientific™ C5000-595W
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 14809820
  • 534.52 SEK / Pack of 100
Estimated Shipment
03-04-2019
Add to basket
Description
Description
SureStop Features
  • Provides a consistent seal independent of user, or the amount of torque applied when closing vial
  • Provides optimal septum compression across the opening of the vial
  • Design allows the positioning of the closure to be consistently “flat” producing a more cylindrical vial profile, which improves autosampler recognition and compatibility
  • As a result of AVCS, when used with our C5000 closure, SureStop becomes the best sealing vial/closure on the market
  • Designed for use with nearly every autosampler on the market
  • When compared to other 2mL vials, SureStop yields the lowest loss to evaporation and the lowest standard deviation in evaporation studies
Specifications
Specifications
Blue; White
2mL, Clear Glass w/ID Patch, with Blue PTFE/White Silicone, Pre-slit Septum
100
Clear Glass
Glass, PTFE, Silicone
PTFE; Silicone
Yes
Blue PTFE/White Silicone, Pre-slit Septum
Clear
2 mL
Videos
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.