Läs mer
(9R)-9-(2-Pyridinyl)-6-oxaspiro[4.5]decane-9-acetonitrile, TRC
Produktkod. 30425636
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425636 500 mg
30414145 250 mg
30419009 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425636 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP996960500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1401031-38-6
Molekylformel C16H20N2O
Molekylvikt (g/mol) 256.34
IUPAC-namn 2-[(9R)-9-pyridin-2-yl-6-oxaspiro[4.5]decan-9-yl]acetonitrile
LEDER N#CC[C@]1(CCOC2(CCCC2)C1)c3ccccn3
Specifikationer
Kemiskt namn eller material (9R)-9-(2-Pyridinyl)-6-oxaspiro[4.5]decane-9-acetonitrile
Kvantitet 500 mg
InChI formel InChI=1S/C16H20N2O/c17-10-8-15(14-5-1-4-11-18-14)9-12-19-16(13-15)6-2-3-7-16/h1,4-5,11H,2-3,6-9,12-13H2/t15-/m1/s1
Formel vikt 256.1576