missing translation for 'onlineSavingsMsg'
Läs mer

(9Z,11E)-Methyl Ester 9,11-Octadecadienoate, TRC

Produktkod. 30009971
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009971 25 mg -
30009119 10 mg -
30009532 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. View return policy
Product Code. 30009971 Supplier Toronto Research Chemicals Supplier No. TRCM32530525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13058-52-1
Molekylformel C19 H34 O2
Molekylvikt (g/mol) 294.47
Synonym Methyl 9-cis-11-trans-octadecadienoate
IUPAC-namn methyl (9Z,11E)-octadeca-9,11-dienoate
LEDER CCCCCC\C=C\C=C/CCCCCCCC(=O)OC

Specifications

Kemiskt namn eller material (9Z,11E)-Linoleic acid methyl ester
Kvantitet 25 mg
InChI formel InChI=1S/C19H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h8-11H,3-7,12-18H2,1-2H3/b9-8+,11-10-
Formel vikt 294.2559
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.