Läs mer
(9Z,12E)-Tetradecadien-1-yl Acetate, TRC
Produktkod. 30137503
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137503 250 mg
30135404 50 mg
17983275 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137503 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT450085250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 31654-77-0
Molekylformel C16 H28 O2
Molekylvikt (g/mol) 252.39
Synonym 9,12-Tetradecadien-1-ol, acetate, (9E,12Z)- (9CI), 9,12-Tetradecadien-1-ol, acetate, (Z,E)- (8CI)
IUPAC-namn [(9E,12Z)-tetradeca-9,12-dienyl] acetate
LEDER C\C=C/C\C=C\CCCCCCCCOC(=O)C
Specifikationer
Kemiskt namn eller material (Z,E)-9,12-Tetradecadien-1-yl acetate
Kvantitet 250 mg
InChI formel InChI=1S/C16H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-18-16(2)17/h3-4,6-7H,5,8-15H2,1-2H3/b4-3-,7-6+
Formel vikt 252.2089