Läs mer
(9Z,12Z)-9,12-Octadecadien-1-ol, TRC
Produktkod. 30418862
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30418862 500 mg
30414761 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30418862 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO848460500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Linoleyl Alcohol
CAS 506-43-4
Molekylformel C18 H34 O
Kvantitet 500 mg
Synonym Linoleyl Alcohol, (9Z,12Z)-octadeca-9,12-dien-1-ol
InChI formel InChI=1S/C18H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h6-7,9-10,19H,2-5,8,11-18H2,1H3/b7-6-,10-9-
LEDER CCCCC\C=C/C\C=C/CCCCCCCCO
IUPAC-namn (9Z,12Z)-octadeca-9,12-dien-1-ol
Molekylvikt (g/mol) 266.46
Formel vikt 266.261