Läs mer
Diiodo(p-cymene)ruthenium(II) dimer, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 046886.06
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifikationer
| Diiodo(p-cymene)ruthenium(II) dimer | |
| MFCD08064027 | |
| Light and moisture sensitive | |
| Insoluble in water. |
| 90614-07-6 | |
| 5g | |
| Di-μ-iodobis[(p-cymene)iodoruthenium(II)] | |
| 978.2 |