Läs mer
Busulfan, 98%, Thermo Scientific™
A cell cycle non-specific alkylating antineoplastic agent
Brand: Thermo Scientific Alfa Aesar J61348.14
| Kvantitet | 25g |
|---|
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifikationer
| Busulfan | |
| C6H14O6S2 | |
| MFCD00007562 | |
| UN2811 | |
| 14,1505 | |
| COVZYZSDYWQREU-UHFFFAOYSA-N | |
| 4-methylsulfonyloxybutyl methanesulfonate | |
| 2478 | |
| 246.29 |
| 55-98-1 | |
| CH3SO2O(CH2)4OSO2CH3 | |
| 25g | |
| 1791786 | |
| busulfan, myleran, busulphan, sulphabutin, leucosulfan, myelosan, busulfex, citosulfan, mielucin, misulban | |
| CS(=O)(=O)OCCCCOS(=O)(=O)C | |
| 246.292 | |
| CHEBI:28901 | |
| 98% |