Läs mer
Tocris Bioscience™ A 1120
GREENER_CHOICE
Produktkod. 18094712
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18094712 10 mg
18005072 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18094712 Leverantör Tocris Bioscience™ Leverantörsnummer 3793/10
High affinity retinol-binding protein 4 (RBP4) ligand
High affinity retinol-binding protein 4 (RBP4) ligand (Ki = 8.3 nM); non-retinoid. Selective against a range of different cellular targets. As efficacious as fenretinide (Cat. No. 1396) in the reduction of serum RBP4 and retinol. Displaces transthyretin (TTR) from RBP4-TTR complexes.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1152782-19-8
Molekylformel C20H19F3N2O3
Specifikationer
Kemiskt namn eller material A 1120
Kvantitet 10 mg
Formel vikt 392.37
Renhet >99%
Molekylvikt (g/mol) 392.378