missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ A 1120
GREENER_CHOICE

Produktkod. p-4817461
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 18094712

Brand: Tocris Bioscience™ 3793/10

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High affinity retinol-binding protein 4 (RBP4) ligand

High affinity retinol-binding protein 4 (RBP4) ligand (Ki = 8.3 nM); non-retinoid. Selective against a range of different cellular targets. As efficacious as fenretinide (Cat. No. 1396) in the reduction of serum RBP4 and retinol. Displaces transthyretin (TTR) from RBP4-TTR complexes.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1152782-19-8
Molekylformel C20H19F3N2O3

Specifica

Kemiskt namn eller material A 1120
Kvantitet 10 mg
Formel vikt 392.37
Renhet >99%
Molekylvikt (g/mol) 392.378
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.