Läs mer
A-395, MedChemExpress
Produktkod. 16483398
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16483398 10 mg
16446127 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16483398 Leverantör Medchem Express Leverantörsnummer HY101512
MedChemExpress A-395 is an antagonist of polycomb repressive complex 2 (PRC2) protein-protein interactions that potently inhibits the trimeric PRC2 complex (EZH2-EED-SUZ12) with an IC50 of 18 nM.
A-395 is an antagonist of polycomb repressive complex 2 (PRC2) protein-protein interactions that potently inhibits the trimeric PRC2 complex (EZH2-EED-SUZ12) with an IC50 of 18 nM.
Specifikationer
Kemiskt namn eller material A-395
CAS 2089148-72-9
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C26H35FN4O2S
Kvantitet 10 mg
Löslighetsinformation DMSO : 100 mg/mL (205.49 mM; Need ultrasonic)
LEDER O=S(N1CCN(C2=CC=C([C@H]3CN(C4C(C(F)=CC=C5)=C5CC4)C[C@@H]3N(C)C)C=C2)CC1)(C)=O
Molekylvikt (g/mol) 486.65
Formel vikt 486.65
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation