missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ A 410099.1 amide-alkylC4-amine
GREENER_CHOICE

Produktkod. 18756346
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
18756346 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18756346 Supplier Tocris Bioscience™ Supplier No. 6474/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

IAP ligand with alkyl linker and terminal amine for onward chemistry

A 410099.1 amide-alkylC4-amine is a functionalized IAP ligand for PROTAC® research and development; incorporates an IAP ligand plus an amide-alkylC4 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material tert-Butyl [-1-[[-2-[(2 S,4 S)-4-(5-Aminopentanamido)-2-[[(R)-1,2,3,4-tetrahydronaphthalen-1-yl]carbamoyl]pyrrolidin-1-yl]-1-cyclohexyl-2-oxoethyl]amino]-1-oxopropan-2-yl](methyl)carbamate
CAS 2415256-18-5
Kvantitet 25 mg
Mål Degrader Building Blocks
Molekylformel C37H58N6O6
Renhet 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.