missing translation for 'onlineSavingsMsg'
Läs mer

A 438079 hydrochloride, MedChemExpress

Produktkod. 16496568
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
Förpackningsstorlek:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16496568 10 mg 10mg
16402667 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16496568 Leverantör Medchem Express Leverantörsnummer HY15488A

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress A 438079 (hydrochloride) is a potent, and selective P2X7 receptor antagonist with pIC50 of 6.9.

A 438079 (hydrochloride) is a potent, and selective P2X7 receptor antagonist with pIC50 of 6.9.

Specifikationer

Kemiskt namn eller material A 438079 hydrochloride
Fysisk form Solid
CAS 899431-18-6
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C13H10Cl3N5
För användning med (applikation) Neuroscience-Neuromodulation
Löslighetsinformation H2O : ≥ 350 mg/mL (1021.57 mM) ∣DMSO : ≥ 100 mg/mL (291.88 mM)
LEDER ClC1=C(Cl)C(C2=NN=NN2CC3=CC=CN=C3)=CC=C1.[H]Cl
Molekylvikt (g/mol) 342.61
Formel vikt 342.61
Procent renhet 99.26%
Kvalitet Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.