Läs mer
Tocris Bioscience™ A 438079 hydrochloride
GREENER_CHOICE
Produktkod. 18015682
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18015682 50 mg
18014842 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18015682 Leverantör Tocris Bioscience™ Leverantörsnummer 2972/50
Competitive P2X7 antagonist
A 438079 hydrochloride is a competitive P2X7 receptor antagonist (pIC50 = 6.9 for the inhibition of Ca2+ influx in the human recombinant P2X7 cell line). Devoid of activity at other P2 receptors (IC50 > 10 μM). Possesses antinociceptive activity in models of neuropathic pain in vivo. Inhibits methamphetamine-induced microglial migration and phagocytosis in vitro.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 899431-18-6
Molekylformel C13H10Cl3N5
Specifikationer
Kemiskt namn eller material A 438079 hydrochloride
Kvantitet 50 mg
Formel vikt 342.61
Renhet >98%
Molekylvikt (g/mol) 342.608