Läs mer
Tocris Bioscience™ A 582941
GREENER_CHOICE
Produktkod. 18085852
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18085852 10 mg
18064482 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18085852 Leverantör Tocris Bioscience™ Leverantörsnummer 4341/10
Partial agonist at α7 nAChR
Selective α7 nAChR partial agonist; exhibits high affinity for both rat and human α7 receptors (Ki values are 10.8 and 16.7 nM respectively). Displays ∽15-fold selectivity for α7 over 5-HT3 receptors (Ki = 150 nM for 5-HT3).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 848591-90-2
Molekylformel C17H20N4
Specifikationer
Kemiskt namn eller material A 582941
Kvantitet 10 mg
Formel vikt 280.37
Renhet >98%
Molekylvikt (g/mol) 280.375