Läs mer
A 77-1726, TRC
Produktkod. 30012268
Change view
Kvantitet:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012268 50 mg
30004312 500 mg
30011309 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012268 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA77172650MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 163451-81-8
Molekylformel C12 H9 F3 N2 O2
Molekylvikt (g/mol) 270.21
Synonym (2Z)-2-Cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]but-2-enamide, Leflunomide Imp. B (EP), (2Z)-2-cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]-2-Butenamide, 2-Butenamide, 2-cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]-, (Z)-, A 1726, A 77-1726, A 771726, Aubagio, Flucyamide, HMR 1726, SU 20, Teriflunomide
IUPAC-namn (Z)-2-cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]but-2-enamide
LEDER C\C(=C(/C#N)\C(=O)Nc1ccc(cc1)C(F)(F)F)\O
Specifikationer
Kemiskt namn eller material (2Z)-2-Cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]but-2-enamide (Teriflunomide)
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C12H9F3N2O2/c1-7(18)10(6-16)11(19)17-9-4-2-8(3-5-9)12(13,14)15/h2-5,18H,1H3,(H,17,19)/b10-7-
Formel vikt 270.0616
Procent renhet >95%