Läs mer
Tocris Bioscience™ A 77636 hydrochloride
GREENER_CHOICE
Produktkod. 18044932
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18044932 10 mg
18024372 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18044932 Leverantör Tocris Bioscience™ Leverantörsnummer 1701/10
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Potent and selective D1-like agonist; orally active
Potent and selective dopamine D1-like receptor agonist (pEC50 values are 8.97 and < 5 for D1-like and D2-like receptors respectively). Displays anti-Parkinsonian activity following oral administration in vivo. Exhibits 11-fold cell type bias over dopamine in a functional assay in U2 cells.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 145307-34-2
Molekylformel C20H28ClNO3
Specifikationer
Kemiskt namn eller material A 77636 hydrochloride
Kvantitet 10 mg
Formel vikt 365.9
Renhet >98%
Molekylvikt (g/mol) 365.898