Läs mer
Tocris Bioscience™ A 779
GREENER_CHOICE
Selective Mas receptor (Ang-(1-7) receptor) antagonist
Brand:  Tocris Bioscience™ 5937/5
Encompass
Produktkod. 15707176
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
A 779 is a selective Mas receptor (Ang-(1-7) receptor) antagonist. Exhibits no significant affinity for AT1 or AT2 receptors at a concentration of 1 μM. Inhibits antidiuretic effect of Ang-(1-7) in water-loaded rats. Also attenuates Monocrotaline-induced pulmonary fibrosis in rats.
Specifikationer
Specifikationer
0.95
Store at -20°C
Observed M.W.: 872.98
C39H60N12O11
159432-28-7
Other Angiotensin Receptors
5 mg
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer