Läs mer
a,a-Dichlorotoluene, SPEX CertiPrep™
Produktkod. 16399187 Handla allt SPEX Certiprep Produkter
Change view
Kvantitet:
1 mL
Produktkod. Kvantitet
16399187 1 mL
1 options
Produktkod. 16399187 Leverantör SPEX Certiprep Leverantörsnummer S1472
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Innehåller
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
a,a-Dichlorotoluene98-87-31000μg/mL
Certifikat
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Uppfyllande
GHS/OSHA Complaint
Specifikationer
Kemiskt namn eller material a,a-Dichlorotoluene
Förpackning 2mL Amber Prescored Glass Ampule
CAS 98-87-3, 67-56-1
CAS Min % 0.1, 99.9
Anteckningar om koncentration eller sammansättning Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylformel C7H6Cl2
För användning med (utrustning) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI-nyckel CAHQGWAXKLQREW-UHFFFAOYSA-N
LEDER C1=CC=C(C=C1)C(Cl)Cl
IUPAC-namn dichloromethylbenzene
Lösningsmedel eller matris Methanol-P&T
Molekylvikt (g/mol) 161.025
PubChem CID 7411
Kvantitet 1 mL
ChEBI CHEBI:82273
Visa mer Visa mindre
Product intended for laboratory use only