Läs mer
a-Aminoacetophenone-d5 Hydrochloride, TRC
Produktkod. 30314188
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30314188 100 mg
1 options
Produktkod. 30314188 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA580087100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha-Aminoacetophenone-d5 Hydrochloride
CAS 1794938-50-3
Molekylformel C8H5D5ClNO
Kvantitet 100 mg
Synonym 2-Amino-1-phenyl-1-ethanone-d5 Hydrochloride, 2-Amino-1-phenylethanone-d5 Hydrochloride, 2-Aminoacetophenone-d5 Hydrochloride, Phenacylamine-d5 Hydrochloride
InChI formel InChI=1S/C8H9NO.ClH/c9-6-8(10)7-4-2-1-3-5-7;/h1-5H,6,9H2;1H/i1D,2D,3D,4D,5D;
LEDER Cl.[2H]c1c([2H])c([2H])c(C(=O)CN)c([2H])c1[2H]
IUPAC-namn 2-amino-1-(2,3,4,5,6-pentadeuteriophenyl)ethanone;hydrochloride
Molekylvikt (g/mol) 176.65
Formel vikt 176.0765