Läs mer
a-D-Mannopyranosyl Azide, TRC
Produktkod. 30180170
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180170 1 mg
30179794 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180170 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM1664601MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51970-29-7
Molekylformel C6H11N3O5
Molekylvikt (g/mol) 205.17
Synonym 1-Azido-alpha-D-mannopyranoside;
LEDER OC[C@H]1O[C@H](N=[N+]=[N-])[C@@H](O)[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material a-D-Mannopyranosyl Azide