Läs mer
a-Desmethyl Anastrozole(>90%), TRC
Produktkod. 30136817
Change view
Kvantitet:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136817 25 mg
18043330 10 mg
18080840 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136817 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29073025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1215780-15-6
Molekylformel C16 H17 N5
Molekylvikt (g/mol) 279.34
Synonym 1,3-Benzenediacetonitrile, α1,α1,α3-trimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-, α1,α1,α3-Trimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile, 2-[3-[(1RS)-1-Cyanoethyl]-5-(1H-1,2,4-triazol-1-ylmethyl)phenyl]-2-methylpropanenitrile
IUPAC-namn 2-[3-(1-cyanoethyl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-2-methylpropanenitrile
LEDER CC(C#N)c1cc(Cn2cncn2)cc(c1)C(C)(C)C#N
Specifikationer
Kemiskt namn eller material alpha1,alpha1,alpha3-Trimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile
Kvantitet 25 mg
InChI formel InChI=1S/C16H17N5/c1-12(7-17)14-4-13(8-21-11-19-10-20-21)5-15(6-14)16(2,3)9-18/h4-6,10-12H,8H2,1-3H3
Formel vikt 279.1484