Läs mer
a-iso-Methylionone, TRC
Produktkod. 30475050
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30475050 500 mg
1 options
Produktkod. 30475050 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM262613500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha-Isomethylionone
CAS 127-51-5
Molekylformel C14 H22 O
Kvantitet 500 mg
Synonym 3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one, 4-(2,6,6-Trimethyl-2-cyclohexen-1-yl)-3-methyl-3-buten-2-one, Cetone Alpha, Isomethyl-α-ionone, NSC 66432, α-Cetone, α-Ionone, isomethyl-
InChI formel InChI=1S/C14H22O/c1-10-7-6-8-14(4,5)13(10)9-11(2)12(3)15/h7,9,13H,6,8H2,1-5H3
LEDER CC(=O)C(=CC1C(=CCCC1(C)C)C)C
IUPAC-namn 3-methyl-4-(2,6,6-trimethylcyclohex-2-en-1-yl)but-3-en-2-one
Molekylvikt (g/mol) 206.32
Formel vikt 206.1671