Läs mer
(-)-a-Kainic Acid, TRC
Produktkod. 30405133
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30405133 10 mg
1 options
Produktkod. 30405133 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK13501010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (-)-alpha-Kainic Acid
CAS 487-79-6
Molekylformel C10 H15 N O4
Kvantitet 10 mg
Synonym (2S,3S,4S)-2-Carboxy-4-(1-methylethenyl)-3-pyrrolidineacetic Acid, 2-Carboxy-4-isopropenyl-3-pyrrolidineacetic Acid, (-)-Kainic Acid, (2S,3S,4S)-2-Carboxy-4-isopropenylpyrrolidine-3-acetic Acid, Digenic Acid, Digenin, Helminal, Kainic Acid, L-α-Kainic Acid, α-Kainic Acid
InChI formel InChI=1S/C10H15NO4/c1-5(2)7-4-11-9(10(14)15)6(7)3-8(12)13/h6-7,9,11H,1,3-4H2,2H3,(H,12,13)(H,14,15)/t6-,7+,9-/m0/s1
LEDER CC([C@H]1CN[C@H](C(O)=O)[C@H]1CC(O)=O)=C
IUPAC-namn (2S,3S,4S)-3-(carboxymethyl)-4-prop-1-en-2-ylpyrrolidine-2-carboxylic acid
Molekylvikt (g/mol) 213.23
Formel vikt 213.1001