missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tocris Bioscience™ (+/-)-a-Lipoamide, Phase Separated
Description
(±)-α-Lipoamide is a phase-separated condensate modifier. Reduces propensity of stress granule proteins to aggregate (EC50 = 20 μM in HeLa and iPSCs), and causes FUS stress granule proteins to return to the nucleus in vitro. Stimulates mitochondrial biogenesis in 3T3-L1 adipocytes. Prevents dieback of ALS patient-derived (FUS mutant) motor neuron axons in culture, and recovers motor defects in D. melanogaster expressing FUS mutants. Suppresses the effects of ALS-associated FUS mutations in vivo in different systems. Orally bioavailable.
Specifications
Specifications
| Kemiskt namn eller material | 1,2-Dithiolane-3-pentanamide |
| CAS | 940-69-2 |
| Kvantitet | 100 mg |
| Molekylformel | C8H15NOS2 |
| Renhet | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?