Läs mer
a-Methyl-L-tyrosine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | a-Methyl-L-p-tyrosine |
| CAS | 672-87-7 |
| Molekylformel | C10 H13 N O3 |
| Kvantitet | 500 mg |
| Synonym | L-Tyrosine, α-methyl-, Tyrosine, α-methyl-, L- (8CI), α-Methyl-L-tyrosine, (S)-2-(4-Hydroxybenzyl)-2-aminopropanoic acid, (S)-α-Methyltyrosine, Demser, L(-)-Metyrosine, L-Metyrosine, L-α-MT, L-α-Methyl-p-tyrosine, L-α-Methyltyrosine, MK 781, Metirosine, Metyrosine |
| InChI formel | InChI=1S/C10H13NO3/c1-10(11,9(13)14)6-7-2-4-8(12)5-3-7/h2-5,12H,6,11H2,1H3,(H,13,14)/t10-/m0/s1 |
| LEDER | C[C@](N)(Cc1ccc(O)cc1)C(=O)O |
| IUPAC-namn | (2S)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid |
| Molekylvikt (g/mol) | 195.22 |
| Formel vikt | 195.0895 |