Läs mer
a-Methyl-trans-cinnamaldehyde, TRC
Produktkod. 30464588
Change view
Kvantitet:
50 g
Produktkod. Kvantitet
30464588 50 g
1 options
Produktkod. 30464588 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30495350G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha-Methyl-trans-cinnamaldehyde
CAS 15174-47-7
Molekylformel C10H10O
Kvantitet 50 g
Synonym 2-Methyl-3-phenyl-2-propenal, 2-Methyl-3-phenyl-2-propen-1-al, 2-Methyl-3-phenylacrolein, 2-Methyl-3-phenylacrylaldehyde, 2-Methyl-3-phenylpropenal, 2-Methylcinnamic Aldehyde, Methylcinnamaldehyde, NSC 22283, NSC 49286, alpha-Methylcinnamaldehyde, alpha-Methylcinnamic Aldehyde, alpha-Methyl-cinnamaldehyde
InChI formel InChI=1S/C10H10O/c1-9(8-11)7-10-5-3-2-4-6-10/h2-8H,1H3/b9-7+
LEDER C\C(=C/c1ccccc1)\C=O
IUPAC-namn (E)-2-methyl-3-phenylprop-2-enal
Molekylvikt (g/mol) 146.19
Formel vikt 146.0732