missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ A4GALT Monoclonal Antibody (38.13), PE

Rat Monoclonal Antibody
Brand: Invitrogen™ MA544106
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody recognizes CD77 (globotriaosylceramide Gb3), a neutral glycosphingolipid expressed mainly on the surface of B cell populations, such as germinal center B cells and Burkitt's lymphoma cells.
The protein encoded by this gene catalyzes the transfer of galactose to lactosylceramide to form globotriaosylceramide, which has been identified as the P(k) antigen of the P blood group system. The encoded protein, which is a type II membrane protein found in the Golgi, is also required for the synthesis of the bacterial verotoxins receptor.
Specifikationer
| A4GALT | |
| Monoclonal | |
| PE | |
| A4galt | |
| A14GALT; A4GALT; A4galt {ECO:0000312; A4GALT1; alpha 1,4-galactosyltransferase; alpha 14-galactosyltransferase; alpha-1,4-galactosyltransferase; Alpha-1,4-N-acetylglucosaminyltransferase; alpha4Gal-T1; cd77; CD77 synthase; Gb3; Gb3 synthase; Gb3/CD77 synthase; Gb3S; Globotriaosylceramide synthase; lac; lactosylceramide 4-alpha-galactosyltransferase; P blood group (P one antigen); P(k); P(k) antigen synthase; P1; P1/Pk synthase; P1PK; PK; RGD:621583}; truncated alpha 1,4-galactosyltransferase; UDP-galactose:beta-D-galactosyl-beta1-R 4-alpha-D-galactosyltransferase | |
| Rat | |
| Size-exclusion chromatography | |
| RUO | |
| 53947 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 38.13 | |
| TBS with 0.2% BSA and 15mM sodium azide; pH 8 | |
| Q9NPC4 | |
| A4galt | |
| Daudi cell line (Burkitts lymphoma). | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering